missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences DURAN™ GLS 80™ Wide Mouth Bottle Connection System, Screw Cap GLS 80™, 4-Ports, GL 18 Thread
Flexible GLS 80™ connection system for the use of semi-rigid hoses.
Supplier: DWK Life Sciences 1160170
Description
Flexible GLS 80™ cap connection system for use with semi-rigid tubing, suitable for DURAN™ GLS 80™ laboratory and media bottles. Five different tube diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation through membrane liter possible. Unused ports can be provided with a blind cap. Exemplary applications: Safe transfer of liquid media within a closed and sterile system.
- The flexible modular system allows individual combination of the various components according to requirements and application
- Five different hose diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected
- NS 29/32 connection in the middle for tight closure
- Sterile pressure equalisation through membrane filter possible
- Unused ports can be fitted with a GL 18 blind cap
- Compatible with all GLS 80 threads and therefore versatile
Specifications
Insert for screw cap GL 18, ID 6.0 mm | |
Polypropylene, PTFE | |
Autoclavable |
Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor | |
1 Ea. | |
87 mm |