missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT
PBT screw cap with an open top and high temperature and chemical resistance.
$102.18 - $103.87
Specifications
| Color | Red |
|---|---|
| For Use With (Application) | Aseptic reagent delivery and culture sampling using a syringe and needle |
| Material | PBT |
| Quantity | 10/Pk. |
| Screw Material | Polybutylene Terephthalate (PBT) with glass fiber |
| Catalog Number | Mfr. No. | Thread Size (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Thread Size (Metric) | Price | Quantity | |||||
|
09-841-253
|
DWK Life Sciences
292270508 |
GL14 |
Pack of 10 Each for $103.87
|
|
|||||
|
09-841-254
|
DWK Life Sciences
292270902 |
GL 25 |
Pack of 10 Each for $102.18
|
|
|||||
Description
Open topped cap with central aperture suitable for use with bromobutyl stoppers, silicone piercing septa or glassware connections. More chemically resistant than PP cap.- Compatible with GL bottle threads
- Red PBT offers high temperature and chemical resistance
- Glass reinforced PBT offers high strength
Specifications
| Red | |
| PBT | |
| Polybutylene Terephthalate (PBT) with glass fiber | |
| PBT with Glass Fiber |
| Aseptic reagent delivery and culture sampling using a syringe and needle | |
| 10/Pk. | |
| Open Top Screw Cap | |
| Autoclavable |