missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences DURAN™ Stainless Steel 3-Port Connector Cap GL 45

Tubing connection system for use with all sizes of DURAN GL 45 laboratory and media bottles. Wheaton™ DURAN™ Stainless-steel 3-Port Connector Cap GL 45 is ideal for chemistry, life science and biopharma laboratories.

Supplier:  DWK Life Sciences 292612804

Catalog No. 14-209-166


Explore available promotions

Description

Description

  • Bottle top adaptor facilitates the transfer of liquids within a closed and sterile system
  • Three barbed, male hose connectors on top and bottom assemblies are suitable for flexible tubing with 8mm inner diameter, such as Sani-Tech™ Ultra (ULTRA-C-312) Silicone tubing
  • Freely rotating stainless steel centre allows bottle to be unscrewed without need to disconnect tubing
  • Three top connectors are slightly splayed to facility easier attachment of tubing
  • Connectors and body are manufactured from Type 316L (1.4404 / S31603) stainless steel
  • Components are fully welded to avoid entrapment voids
  • Electropolished surface for efficient cleaning and sterilization
  • Liquid tight seal is provided by a silicone gasket and the red PBT GL 45 screw cap
  • Temperature usage range (up to 180°C)
  • Autoclavable (at 121°C / 15 minutes)
  • Retrace coded for full traceability
Applications:
  • Sterile liquid transfer between containers
  • Liquid sampling into bottles from larger containers
  • Reservoir cap for supplying laboratory instruments
  • Port assemblies for spinner flasks, and cell culture vessels
Specifications

Specifications

Stainless Steel
Autoclavable
Connector Cap (3-port)
GL 25
Videos
SDS
Documents

Documents

Promotions

Promotions