missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynabeads™ Protein A IP Kit and Magnet Starter Pack
The Dynabeads Protein A IP Kit and Magnet Starter Pack combines Dynabeads Protein A magnetic beads for immunoprecipitation (including the required buffers) and the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience.
Supplier: Invitrogen™ 10018D
Description
Space-saving design with our smallest thermal cycler footprint
- Flexible modular design saves space on deck
- The separate control panel module provides flexibility when integrating on the robotic deck
- Choice of 10 cm, 1 meter and 3 meter cabling options
Automated lid and cycler features
- 384-well block
- The automated lid is compatible with programming for easy hands-free operation with liquid handling or plate stackers
- The heated cover slides forward to cover the plate nest,ensuring thermal contact and eliminating evaporation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
| 2 mL Dynabeads Protein A beads; 16 mL Antibody Binding and Washing Buffer; 28 mL Washing Buffer; 1 mL Elution Buffer; 1 x DynaMag-2 Magnet | |
| Room Temperature | |
| 40 reactions | |
| IP Kit and Magnet Starter Pack |
| Store Dynabeads at 2–8°C. | |
| DynaMag, Dynabeads | |
| Immunoprecipitation | |
| Protein A |
For Research Use Only. Not for use in diagnostic procedures.