Learn More
Invitrogen™ DynaMag™-96 Side Skirted Magnet
For use with Dynabeads ranging from 1–4.5 μm in diameter. Features six bar magnets and is compatible with most 96-well skirted PCR-plates, culture plates, 96-well round bottom plates, 96-well flat bottom plates, 24-well plates, 12-well plates, and 6-well plates.
Supplier: Invitrogen™ 12027
Includes: one magnet
Description
For use with Dynabeads ranging from 1–4.5 μm in diameter. Features six bar magnets and is compatible with most 96-well skirted PCR-plates, culture plates, 96-well round bottom plates, 96-well flat bottom plates, 24-well plates, 12-well plates, and 6-well plates. Working volume per well (5–200 μL).
- Efficient mixing without sample handling
- Mixing can be performed by shifting the 96-well plate back and forth from right-most position to left-most position
- Shifting draws beads back and forth through the sample and allows mixing to be achieved without handling, promoting faster processing and less sample handing-induced variation
- When used in combination with DynaMag-96 Bottom magnet on automation platform, two magnets allow for significant volume changes through a workflow
- High wash volumes: DynaMag-96 Side magnet and DynaMag-96 Side Skirted magnet
- Low elution volumes: DynaMag-96 Bottom magnet
Plates size may vary. Test your plate for compatibility. Not recommended for use with 48-well plates.
Order Info
Shipping Condition: Room Temperature
Specifications
| Magnet | |
| DynaMag™ | |
| DYNAL, Dynabeads, MPC |
| Contains one magnet. Store at room temperature. | |
| Room Temperature | |
| 1 Each |
For Research Use Only. Not for use in diagnostic procedures.