missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaMag™-2 Magnet DFS Item

The DynaMag-2 magnet combines a strong magnetic attraction for efficient separations in standard microcentrifuge tubes with a flexible and smart ergonomic design.

Supplier:  Invitrogen™ 12321D

Catalog No. 12321D


Only null left
Explore available promotions

Includes: One DynaMag-2 magnet

Description

Description

The DynaMag-2 magnet combines a strong magnetic attraction for efficient separations in standard microcentrifuge tubes with a flexible and smart ergonomic design.

  • Optimal working volume: 10–2000 μL
  • Holds up to 16 standard 1.5–2 mL microcentrifuge tubes in numbered spaces
  • Good control and visibility of sample
  • Top rack can be quickly removed from magnet in base, ready for resuspension, vortexing, rotation, or manual sample shaking
  • Magnet can be used with all types of Dynabeads for efficient isolation of proteins and nucleic acids
  • Sample rack can be easily removed from base magnet for resuspension, vortexing, rotation or manual sample shaking
  • Center pin in rack ensures equal vortexing of all tubes
  • Rack can also be removed and used to store tubes
  • 16 numbered sample spaces make magnet for parallel handling of multiple samples
  • With eight tube positions on each side of rack, easily scale protocols to accommodate 96-well plate reactions

This magnet is not compatible with the MX4 Mixer. The removable sample rack is also available as a separate item. Purchasing an additional sample rack minimizes the need to move tubes between racks. The rack can be placed on the bench, in the refrigerator, or in the incubator.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Magnet
Room Temperature
1 Each
Contains one DynaMag™-2 magnet. Store at room temperature.
DYNAL, DynaMag, Dynabeads
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.