missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ DynaLight™ Substrate with RapidGlow™ Enhancer
A ready-to-use chemiluminescent substrate formulation optimized to achieve faster results in solution-based immunoassays. Thermo Scientific DynaLight Substrate with RapidGlow Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline
$956.00 - $956.00
Specifications
Product Type | ELISA Substrate |
---|---|
Detection Method | Chemiluminescence |
Assay Sensitivity | 1 pg/mL |
Form | Liquid |
Format | Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
4475406
|
Invitrogen™
4475406 |
100 mL |
Each for $956.00
|
|
|||||
Description
When combined with DynaLight Trigger Solution, DynaLight Substrate with RapidGlow Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.
- Faster kinetics and higher signal levels than colorimetric, fluorescent, or other chemiluminescent chemistries
- Sensitive attomole detection of purified alkaline phosphatase enzyme
- Optimized formulation for both microplate and bead-based assays
- Substrate stability of over 1 year at 4°C
- Compatibility with automated platforms with trigger solution/positions already in place
- Includes 1,2-dioxetane chemiluminescent substrate and a polymeric enhancer that enables ultra-sensitive immunoassay detection by alkaline phosphatase label
Specifications
ELISA Substrate | |
1 pg/mL | |
Bottle | |
High-throughput Compatible | |
Cell Cultures, Whole Blood, Serum | |
Alkaline Phosphatase | |
1 amber bottle — Store at 2 to 8°C |
Chemiluminescence | |
Liquid | |
Luminometer (Microplate) | |
DynaLight™, NovaBright™, RapidGlow™ | |
Chemical Substrate | |
2 min. |
For Research Use Only. Not for human or animal therapeutic or diagnostic use.