missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dynalon™ Bio-Bin™ Waste Disposal Container Stands
Accessories for Dynalon Bio-Bin Waste Disposal Containers
Description
For use with Dynalon Bio-Bin Waste Disposal Containers (Cat. No. 12—009—800)
Specifications
UN 3291 | |
31.29 cm | |
U Stand | |
2 x 2 in. biohazard waste labels, four (4) non slip feet | |
6.0L Bio-Bin Waste Container | |
7.1 in. | |
Stainless Steel |
7.1 x 10.8 x 12.32 in. (18.03 x 27.43 x 31.29 cm) | |
18.03 cm | |
27.43 cm | |
U stand holds the 6L Bio-bin at a 30° angle, making it ideal for use in a fume cabinet, biological safety cabinet, other workstation or bench | |
10.8 in. | |
12.32 in. | |
Waste Container Stand |