missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dynalon™ Bio-Bin™ Waste Disposal Container Stands

Accessories for Dynalon Bio-Bin Waste Disposal Containers

 View more versions of this product

Catalog No. 12-009-804


Explore available promotions

Description

Description

For use with Dynalon Bio-Bin Waste Disposal Containers (Cat. No. 12—009—800)

Specifications

Specifications

UN 3291
19.9 cm
Z Stand
2 x 2 in. biohazard waste labels, four (4) non slip feet
6.0L Bio-Bin Waste Container
6.7 in.
Stainless Steel
6.7 x 6.7 x 7.87 in. (17.01 x 17.08 x 19.9 cm)
17.01 cm
17.08 cm
Z stand holds the 6L Bio-bin 6 upright, either for bench or floor use and allows for easy removal of used bins
6.7 in.
7.87 in.
Waste Container Stand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions