missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dynamic Diagnostics Paper Towel Dispensers DFS Item

Durable acrylic dispenser holds virtually any size tri-fold or c-fold paper towel

$162.53 - $162.53

Specifications

Product Type Tri-fold Paper Towel Dispenser
Length (English) 11 in.
Length (Metric) 279.4 mm
Material Acrylic
For Use With (Application) Protects towels from moisture and contamination
View More Specs

Products 1
Catalog Number Mfr. No. Dimensions (L x W x H) Size Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Size Price Quantity  
22-899-014 DFS Item
View Documents
Dynamic Diagnostics
400014
11 x 4.5 x 6 in. Small
Each for $162.53
 
Description

Description

  • Keeps workspaces neat and tidy
  • Protects towels from moisture and contamination
  • May be wall mounted using the pre-drilled holes or placed on the countertop
  • Front access allows user to take as many towels as needed at once

Specifications

Specifications

Tri-fold Paper Towel Dispenser
279.4 mm
Protects towels from moisture and contamination
11 in.
Acrylic
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'