missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dysprosium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 040574NA

 View more versions of this product

Catalog No. AA40574NA


Explore available promotions

Description

Description

A solution of 2,6-diformyl-4-methylphenol (0.15mmol) and benzoylhydrazide (0.30 mmol) in MeOH (10 ml) was allowed to react with the dysprosium(III) perchlorate hexahydrate (0.3 mmol) in 10 ml MeOH In observation of slow magnetic relaxation in triple-stranded lanthanide helicates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dysprosium(III) perchlorate hexahydrate
Liquid
Cl3DyO12
UN3211
Insoluble in water.
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Dy+3]
460.838
568.94 (460.85 Anhydrous)
50% w/w aq. soln.
14692-17-2
2 g
MFCD00150227
dysprosium 3+ perchlorate, dysprosium iii perchlorate, dysprosium perchlorate, acmc-1c5o7, dysprosium iii perchlorate solution, perchloric acid dysprosium iii salt, dysprosium 3+ triperchlorate, perchloric acid,dysprosium 3+ salt 8ci,9ci, perchloric acid,dysprosium 3+ salt, hexahydrate 8ci,9ci
QNDHOIIJUAAORW-UHFFFAOYSA-K
dysprosium(3+);triperchlorate
13767046
Reagent
Odorless

Chemical Identifiers

14692-17-2
460.838
QNDHOIIJUAAORW-UHFFFAOYSA-K
13767046
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Dy+3]
Cl3DyO12
MFCD00150227
dysprosium 3+ perchlorate, dysprosium iii perchlorate, dysprosium perchlorate, acmc-1c5o7, dysprosium iii perchlorate solution, perchloric acid dysprosium iii salt, dysprosium 3+ triperchlorate, perchloric acid,dysprosium 3+ salt 8ci,9ci, perchloric acid,dysprosium 3+ salt, hexahydrate 8ci,9ci
dysprosium(3+);triperchlorate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H271-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.

EINECSNumber : 237-838-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only