missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dysprosium ingot, 99.8% (REO), Thermo Scientific Chemicals
$215.49 - $628.21
Chemical Identifiers
CAS | 7429-91-6 |
---|---|
Molecular Formula | Dy |
Molecular Weight (g/mol) | 162.50 |
MDL Number | MFCD00010982 |
InChI Key | KBQHZAAAGSGFKK-UHFFFAOYSA-N |
Synonym | ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n |
PubChem CID | 23912 |
ChEBI | CHEBI:33377 |
IUPAC Name | dysprosium |
SMILES | [Dy] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged in air stable poly bottle
Chemical Identifiers
7429-91-6 | |
162.50 | |
KBQHZAAAGSGFKK-UHFFFAOYSA-N | |
23912 | |
dysprosium |
Dy | |
MFCD00010982 | |
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n | |
CHEBI:33377 | |
[Dy] |
Specifications
7429-91-6 | |
MFCD00010982 | |
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n | |
[Dy] | |
162.50 | |
CHEBI:33377 | |
Odorless | |
Ingot | |
(REO) |
Dy | |
25 g | |
KBQHZAAAGSGFKK-UHFFFAOYSA-N | |
dysprosium | |
23912 | |
99.8% | |
Poly bottle | |
Dysprosium ingot |
Safety and Handling
EINECSNumber : 231-073-9
TSCA : Yes
storageNote1 :
- Packaged in air stable poly bottle
Recommended Storage : Ambient temperatures
RUO – Research Use Only