missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dysprosium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals

$202.73 - $952.63

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1289706
View Documents
Thermo Scientific Chemicals
01289706
5 g
Each for $202.73
 
AA1289714
View Documents
Thermo Scientific Chemicals
01289714
25 g
Each for $952.63
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7429-91-6
162.50
KBQHZAAAGSGFKK-UHFFFAOYSA-N
23912
dysprosium
Dy
MFCD00010982
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
CHEBI:33377
[Dy]

Specifications

Silver
Dy
UN3089
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
[Dy]
162.50
CHEBI:33377
−40 Mesh
Air and moisture sensitive
Dysprosium powder
7429-91-6
MFCD00010982
5 g
KBQHZAAAGSGFKK-UHFFFAOYSA-N
dysprosium
23912
99.9%
Odorless
Powder
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

EINECSNumber : 231-073-9

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only