Eagle™ Stainless-Steel Safety Cans
The Eagle Stainless-Steel Safety Can is ideal for storage of high-purity liquids and flammables that do not corrode metal
Supplier:  Eagle™ 1301
 View more versions of this product
Catalog No. 04-664-5
Explore available promotions
Description
Description
Stainless-steel mesh flame arrester dissipates heat and flame, preventing ignition of flammable vapors.
  • Dent and puncture resistant
  • Guard cap protects spout and prevents spills if can is dropped
  • Convenient, free-swinging safe handle facilitates carrying and pouring
  • Shock rim protects bottom from abrasion wear and also acts as grip when pouring
  • One electrically welded top-to-bottom seam reduces the possibility of leaks if the can is bumped or dropped
Disposal Cans
  • For intermediate storage of flammable liquid wastes
  • Large 4 in. diameter (10.2cm) spout with lock-open cap facilitates filling and emptying
  • Automatic pressure release opens cap assembly to prevent dangerous buildup of vapors
Safety Cans
  • Heavy-duty protection for temporary storage of flammable liquids
  • Choose standard round or space-saving oblong design
Bottom-Faucet Safety Can
Convenient self-closing bottom faucet enables safe dispensing of flammable liquids into smaller containers from a workbench or elevated shelf
Certifications
UL listed, FM approved.
Compliance
Meet OSHA requirements.
Specifications
Specifications
Oblong Lab Safety Can
Electric Seam Weld
3.8 L
For intermediate storage of flammable liquid wastes
Mesh flame arrester, guard cap, electrically-welded seam
6 in.
13 in.
3.75 lb.
FM, OSHA 29 CFR 1910.106, 1925.152, NFPA Code 30
1 gal.
Stainless Steel
Stainless Steel
8 in.
152 mm
330 mm
1.7 Kg