Thermo Scientific™ EASY-Column™ Capillary HPLC Column Connector Kits
Use these connectors, tees and sleeves with Thermo Scientific™ EASY-Column™ Capillary HPLC Columns.
$203.59 - $712.73
Specifications
Diameter (Metric) 0.79 mm
For Use With LC/MS
Pore Size 120 Å
Product Line EASY-Column
Endcapped Yes
View More Specs
Products 4
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
03-052-613
View Documents Promotion Details
Thermo Scientific™
SC603
Kit for 360μm OD fused silica
Pack of 30 Each for $203.59
03-052-614
View Documents
Thermo Scientific™
SC602
Two-column setup
Each for $712.73
03-052-615
View Documents
Thermo Scientific™
SC601
A/B mixing and venting; for two-column setup
Each for $628.23
03-052-617
View Documents Promotion Details
Thermo Scientific™
SC600
HPLC to column
Each for $298.13
Description
Description
Use these connector kits, sleeves, and tees with Thermo Scientific™ EASY-Column™ Capillary Columns. EASY-Columns optimize nanoscale LC and LC/MS with. Easy-to-use, zero dead volume connector kits are available for one- and two-column configurations and ensure simple, reproducible setup for on-line LC/MS. Each column delivers excellent chromatographic performance on any nanoscale LC system. At flow rates of nL/min versus mL/min, the EASY-Column offers higher sensitivity with greater signal-to-noise ratio compared to traditional electrospray.
Specifications
Specifications
0.79 mm
120 Å
Yes
LC/MS
EASY-Column
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.