Research Products Easy Dip System, Staining Jar, Green, 6 per Package DFS Item
Supplier:  Research Products Corporation 247835G
Unique slide staining system features a square staining jar and a vertical slide rack. Both components are made from heavy duty acetal polymer resistant to alcohol and Xylene. Staining jar has a wide stable base and a recessed interior cavity with a reduced volume capacity of 80ml. Slide staining rack holds up to 12 slides with dimensions of 25 x 75mm, 25x76mm or 1" x 3". Slides are held vertically for easy insertion and removal. Rack is designed for quick drying of slides in a microwave oven or at temperatures up to 120?C. Jars available in five different colors for sample identification. Slide staining rack is sold separately.
Catalog No. 50-999-214
Explore available promotions