Learn More
Thermo Scientific™ EASY-Spray™ HPLC Columns
Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.
$2,675.04 - $4,150.11
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Particle Size | Packing Material | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Particle Size | Packing Material | Price | Quantity | |||
03-251-871
|
Thermo Scientific™
ES900 |
150 mm | 75 μm | 3 μm | Spherical, Fully Porous, Ultrapure Silica |
N/A
|
|
|||
03-251-872
|
Thermo Scientific™
ES901 |
150 mm | 50 μm | 2 μm | Spherical, Fully Porous, Ultrapure Silica |
N/A
|
|
|||
03-251-873
|
Thermo Scientific™
ES902 |
250 mm | 75 μm | 2 μm | Spherical, Fully Porous, Ultrapure Silica |
N/A
|
|
|||
03-251-874
|
Thermo Scientific™
ES903 |
500 mm | 75 μm | 2 μm | Spherical, Fully Porous, Ultrapure Silica |
N/A
|
|
|||
03-251-875
|
Thermo Scientific™
ES904 |
150 mm | 75 μm | 2 μm | Spherical, Fully Porous, Ultrapure Silica |
N/A
|
|
|||
03-251-876
|
Thermo Scientific™
ES905 |
750 mm | 75 μm | 2 μm | Spherical, Fully Porous, Ultrapure Silica |
N/A
|
|
|||
03-251-877
|
Thermo Scientific™
ES906 |
150 mm | 150 μm | 2 μm | Spherical, Fully Porous, Ultrapure Silica |
N/A
|
|
|||
03-251-879
|
Thermo Scientific™
ES911 |
150 mm | 75 μm | 2.6 μm | Spherical, Solid Core, Ultrapure Silica |
N/A
|
|
|||
03-254-131
|
Thermo Scientific™
ES907 |
150 mm | 150 μm | 4 μm | PSDVB (Polystyrene Divinylbenzene) |
N/A
|
|
|||
Description
Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.
EASY-Spray ̶ Fully integrated HPLC column/temperature/emitter design
- Integrated fused-silica emitter delivers an exceptionally stable spray
- Integrated column with temperature control provides maximum reliability and performance
- A single nanoViper connection between the HPLC column and the MS ion source eliminates dead volumes
- Thermo Scientific™ nanoViper™ fingertight fitting prevents overtightening and bad connections
Easy Spray Design Benefits
- Uncompromised performance and ease-of-use in nano LC-MS
- Ultra-sharp peaks
- Outstanding peak capacity
- Maximized peptide coverage
- Outstanding column-to-column reproducibility and reliability
- Outstanding peak capacity for comprehensive proteome characterization
Specifications
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.