EcoLite(+) Scintillation Cocktail, MP Biomedicals
All Purpose, Environmentally Safe, Biodegradable, Non-Toxic, Non-Flammable, Ready-to-Use, For RIA and Receptor Assays, Forms no gel or phasing
Supplier: MP Biomedicals Inc 0188247505
Specifications
EcoLite(+)™ Liquid Scintillation Cocktail | |
5 gal. |