missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edetic Acid, NF, 98-100.5%, Spectrum™ Chemical
E1208, 60-00-4, C10H16N2O8
$11386.20 - $11386.20
Chemical Identifiers
| Molecular Formula | C10H16N2O8 |
|---|---|
| Molecular Weight (g/mol) | 292.24 |
| MDL Number | MFCD00003541 |
| InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
| IUPAC Name | 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid |
| SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602395
|
Spectrum Chemical Mfg Cor
E120825KGBL |
25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Edetic Acid, NF is used as a chelating agent in therapy and as preservative adjuncts in pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| C10H16N2O8 | |
| MFCD00003541 | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid |
| 292.24 | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specifications
| 100% | |
| Fiber Drum | |
| MFCD00003541 | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid | |
| 98 to 100.5% |
| 0.2% | |
| C10H16N2O8 | |
| 25 kg | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
| 292.24 | |
| NF |