missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA, Certified, 0.0575M ±0.0005M (0.115N), LabChem™
$86.60 - $465.70
Chemical Identifiers
| CAS | 6381-92-6 |
|---|---|
| Molecular Formula | C10H18N2Na2O10 |
| Molecular Weight (g/mol) | 372.24 |
| MDL Number | MFCD00150037,MFCD00003541 |
| InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
| Synonym | edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs |
| PubChem CID | 44120005 |
| IUPAC Name | disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
| SMILES | O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
LC138802
|
LabChem
LC138802 |
1 L | Poly Bottle |
Each for $86.60
|
|
||||
|
LC138804
|
LabChem
LC138804 |
4 L | Poly Bottle |
Each for $148.13
|
|
||||
|
LC138806
|
LabChem
LC138806 |
10 L | Cubitainer™ |
Each for $368.50
|
|
||||
|
LC138805
|
LabChem
LC138805 |
20 L | Cubitainer™ |
Each for $465.70
|
|
||||
Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| 44120005 | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
Specifications
| 6381-92-6,7732-18-5 | |
| Colorless | |
| Poly Bottle | |
| (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O | |
| 1 L | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 44120005 | |
| Certified | |
| Liquid |
| 97.86,2.14 | |
| 1g/mL | |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| 1g/mL | |
| Soluble in water | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| 372.23 | |
| 0.0575M ±0.0005M (0.115N) | |
| EDTA |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature