EDTA, Titrant for Water Hardness, (1mL=1mg CaCO3), 0.02N, LabChem™
Supplier:  LabChem LC138501
 View more versions of this product
Catalog No. LC138501
Explore available promotions
Description
Description
Specifications
Specifications
Clear, colorless
Liquid
500 mL
EDTA
0.01000M ±0.00010M (0.02N)
Poly Bottle
Room Temperature