EDTA, Titrant for Water Hardness, (1mL=1mg CaCO3), 0.02N, LabChem™
Supplier:  LabChem LC138504
 View more versions of this product
Catalog No. LC138504
Explore available promotions
Description
Description
Specifications
Specifications
Liquid
4 L
EDTA, 0.01M (0.02N)
PVC Coated
Room Temperature