missing translation for 'onlineSavingsMsg'
Learn More

Eflornithine Hydrochloride Monohydrate 98.0+%, TCI America™

Supplier:  TCI America E0947200MG

Catalog No. E0947200MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Eflornithine Hydrochloride Monohydrate
183°C
C6H15ClF2N2O3
200 mg
FJPAMFNRCFEGSD-UHFFFAOYNA-N
hydrogen 2,5-diamino-2-(difluoromethyl)pentanoic acid hydrate chloride
441361
≥98.0% (T)
96020-91-6
White
MFCD00242734
DL-alpha-Difluoromethylornithine Hydrochloride
[H+].O.[Cl-].NCCCC(N)(C(F)F)C(O)=O
236.64
218.63
Crystalline Powder

Chemical Identifiers

96020-91-6
236.64
FJPAMFNRCFEGSD-UHFFFAOYNA-N
441361
[H+].O.[Cl-].NCCCC(N)(C(F)F)C(O)=O
C6H15ClF2N2O3
MFCD00242734
DL-alpha-Difluoromethylornithine Hydrochloride
hydrogen 2,5-diamino-2-(difluoromethyl)pentanoic acid hydrate chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : RM2981900

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions