missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eicosanedioic Acid 98.0+%, TCI America™

Supplier:  TCI America E0320250G

 View more versions of this product

Catalog No. E0320250G


Explore available promotions

Description

Description

Specifications

Specifications

Eicosanedioic Acid
127°C
234°C
MFCD00059627
eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O
342.52
CHEBI:73728
≥98.0% (T)
2424-92-2
White
C20H38O4
250 g
JJOJFIHJIRWASH-UHFFFAOYSA-N
icosanedioic acid
75502
342.52
Crystalline Powder

Chemical Identifiers

2424-92-2
342.52
JJOJFIHJIRWASH-UHFFFAOYSA-N
75502
icosanedioic acid
C20H38O4
MFCD00059627
eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
CHEBI:73728
C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3083

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions