missing translation for 'onlineSavingsMsg'
Learn More

Bulbtronics™ EIKO Fluorescent Bulb: Shape T5 DFS Item

$13.37 - $13.37

Specifications

Base Style Mini Bi-Pin (G5)
Light Source Fluorescent
Color Blacklight Blue
Diameter (English) 0.62 in.
Diameter (Metric) 1.58 cm
View More Specs

Products 1
Catalog Number Mfr. No. Length (English) Lighting Life Wattage Price Quantity  
Catalog Number Mfr. No. Length (English) Lighting Life Wattage Price Quantity  
14395819 DFS Item
View Documents
Bulbtronics™
17382
6 in. 6000 hr. 4 W
Each for $13.37
Only null left
 
Description

Description

Specifications

Specifications

Mini Bi-Pin (G5)
Blacklight Blue
1.58 cm
T5
Fluorescent
0.62 in.
Stratoliner model 1800, gene splicing equipment
Linear Bulb
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'