missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Demonstration Electric Motor
The permanent magnetic field is provided by the removable bar magnet using this Demonstration Electric Motor.
Supplier: Eisco™ PH1238
Description
- Form of DC electric motor having 2-pole armature wound with enamelled copper wire.
- Disc-type commutator incorporated.
- External connection to phosphor bronze bushes by means of pair of 4 mm sockets.
- Operates on 6 to 8 VDC.
- Dimensions (L x W x H): 5.5 x 5.5 x 7 in. (14 x 14 x 18 cm)
Specifications
6/8 VDC | |
Electric Motor | |
5.5 in. | |
5.51 in. | |
7 in. | |
1.6 lb. |
2-pole armature wound with enameled Copper wire, removable bar magnet, disc-type commutator, external connection to Phosphor Bronze bushes | |
5.5 x 5.51 x 7 in. (14 x 14 x 18 cm) | |
14 cm | |
14 cm | |
18 cm | |
0.71 kg |