Eisco™ Economy Sling Psychrometer Kit
Easy to assemble kit to determine relative humidity.
Supplier: Eisco™ PH1120142PK15
Includes: One dry bulb thermometer, one wet bulb thermometer, one wooden handle, one moisture wick cloth, one screw, two rubber bands, set of instructions with psychometric chart of relative humidity
Description
- Attach the two thermometers to the end of the handle and place the moisture cloth over the wet bulb to assemble
- Just swing the apparatus and then read the difference in temperature between the web-bulb and dry-bulb to determine the relative humidity
- After a few minutes spinning the apparatus the user can take the temperature readings and review the psychometric chart to determine relative humidity
Specifications
One dry bulb thermometer, one wet bulb thermometer, one wooden handle, one moisture wick cloth, one screw, two rubber bands, set of instructions with psychometric chart of relative humidity | |
Economy Sling Psychrometer Kit | |
Determine relative humidity | |
15 Pack |