Eisco™ Heat System Physics Kit
Use the Heat System Physics Kit to explore experiments such as Newton's Law of Cooling, Specific Heat, and the Expansion of Solids, Liquids and Gases.
Supplier:  Eisco™ EIRQ03
Catalog No. S13466
Explore available promotions
Description
Description
  • Over 40 components.
  • Use to teach 15 different Heat activities.
  • Includes Activity Guides with Student worksheets.
  • Packed in Aluminum case with foam cut outs.
Activities Covered
  • Psychrometry
  • The Hot Tea Problem: Newton's Law of Cooling
  • Specific Heat of Several Metals
  • Specific Heat of Water
  • Expansion of Water
  • Heat of Fusion
  • Linear Expansion of a Solid
  • Phase Transitions and Cooling Curves
  • Heat Conduction in a Rod
  • Convective Heat Transfer
  • Pulse Glass
  • Expansion of Gas
  • Expansion of Liquid
  • The Thermocouple
  • The Thermostat
Includes:
Beaker 500 mL (1), Beaker 250 mL (1), Beaker 100 mL (1), Cylinder graduated 100 mL (1), Centrifuge tube 100 mL (1), Filtering flask, with side tube, 500 mL (1), U-shaped glass tube (1), Thermal insulation jacket (1), U-shaped rods, Aluminum (3), U-shaped rods, Brass (3), U-shaped rods, Stainless Steel (3), Pulse glass (1), Glass tube (1), Stand (1), Plastic head with knobs (3), Thermocouple (1), Silicon tubing (1), Extension clamp with rod (2), Alcohol thermometer -10°C to +110°C, (2), Rubber stopper for Filtering flask, Solid (1), Rubber stopper for Filtering flask, one hole (1), Gas burner tubing (1), Bimetallic strip (1), Digital multimeter (1), Calorimeter cup with lid (1), Banana plug cord (2), Silicone grease (1), Paraffin block (4), Stainless Steel and Aluminum cylinders for linear thermal expansion (2), Retort Ring (1), Specific heat cylinders - iron, brass, lead, and copper (4), Wire mesh (1), Bunsen burner (1), Tripod stand for Bunsen burner (1), Micrometer dial gauge range, 0 to 10 mm, sensitivity 0.01 mm (1), Food color (1)
Specifications
Specifications
Heat System Physics Kit
8 to Undergraduate
3 Beakers, 1 Centrifuge Tube, 1 Cylinder, 1 Flask, 1 U-shaped Glass Tube, 1 Long Glass Tube, 9 U-shaped Rods, 1 Pulse Glass, 1 Stand, 3 Plastic Heads, 2 Extension Clamps, 1 Ring, 1 Thermocouple, 1 Silicone Tubing, 1 Thermometer, 2 Rubber Stoppers, 1 Neoprene Tubing, 1 Bunsen Burner, 4 Specific Heat Cylinders, 1 wire Mesh and Tripod Stand, 1 Micrometer Dial Gauge, 1 Food Coloring, 1 Bimetallic Strip, 1 Multimeter, 2 Banana Plug Cords, 1 Calorimeter Cup and Lid, 1 Silicone Grease, 1 Paraffin Wax, 2 Metal Rods, 1 Insulation Jacket