Eisco™ New Calorimeter
Double wall calorimeter of superior quality.
Supplier:  Eisco™ PH0454
Catalog No. S05831
Explore available promotions
Description
Description
  • Easily removable spring heating element unit
  • Operates on 3-6V DC 5w
  • Consists of inner aluminum reservoir 150mL capacity, aluminum outer, plastic insulator ring and styrofoam lining to minimize heat loss
  • Lid accepts heating element, stirrer and thermometer cork includes cover cap
Specifications
Specifications
Double Wall Calorimeter
18 cm
3/6 V
12 cm
7.08 in.
Easily removable spring heating element unit, 150mL inner aluminum reservoir, aluminum outer, plastic insulator ring and styrofoam™ lining, transparent lid with holes, stirrer and built-in thermometer cork and cap
4.72 in.