Learn More
Eisco™ Basic Organic Chemistry Molecular Model Set, 114 Pieces
The 114 Piece Basic Organic Chemistry Molecular Model Set lets students visualize three-dimensional molecule structures and understand molecular formulas.
Supplier: Eisco™ SET00603
Description
- Hard plastic balls of different colors represent atoms
- Each has holes at correct angles to be connected with flexible plastic bonds
- Medium links represent single covalent bonds while long links can be used for double and triple covalent bonds
- Includes the following: 12 Black carbon (4 holes), 20 White hydrogen (1 hole), Six red oxygen (2 holes), Four blue nitrogen (4 holes), Two yellow sulphur (6 holes; 1 hole), One purple phosphorus (4 holes), One gray metal (1 hole), 28 mm Gray links (26), 43 mm Gray links (12), 28 mm White links (26), Assembly tool
Specifications
| Molecular Model Set | |
| Basic Organic Chemistry | |
| 5 cm | |
| 11 in. | |
| ABS Plastic | |
| 1.22 kg | |
| 20 cm |
| 11 x 7.8 x 1.9 in. (28 x 20 x 5 cm) | |
| 1.9 in. | |
| 114 Pc. | |
| 28 cm | |
| 2.68 lb. | |
| 7.8 in. |