Thermo Scientific™ Electroporation Cuvettes
Increase the ease of sample identification and labelling with these premium-grade polycarbonate electroporation cuvettes, which have frosted, colored caps.
Supplier: Thermo Scientific™ MB5520
Description
Increase the ease of sample identification and labeling with premium-grade polycarbonate Thermo Scientific™ Electroporation Cuvettes, which have frosted, colored caps.
- Available in 1mm, 2mm, or 4mm gap sizes
- Pre-sterilized and individually wrapped
- Compatible with all common electroporators
Specifications
Sterile | |
Electroporation Cuvet | |
2mm gap |