Supertek™ Electroscope, Metal Case DFS Item
Enclosed to prevent air currents from affecting the deflection of leaves. Each leaf is suspended from two small hooks.
Supplier:  Supertek™ PH45263
Catalog No. S012322
Explore available promotions
Description
Description
  • Electrosope is enclosed to prevent air currents from affecting the deflection of leaves.
  • Each leaf is suspended from two small hooks.
  • The leaves pivot, rather than bend, when charged.
  • The front and rear side of the box is made of square glass to allow visibility.
  • Supplied with a booklet of 6 leaves
Specifications
Specifications
Electroscope Metal Case with Foil