GSC Go Science Crazy Electroscope Metal Case with Foil
The Electroscope is used for detecting the electrical potential of an object, allowing you to see the presence or absence of an electrical charge.
Supplier:  GSC Go Science Crazy 450170
Catalog No. S13678
Explore available promotions
Alert:
Not a toy. Product intended for ages 13 and up. May contain latex.
Includes: Instructions
Description
Description
Features a glass window box for viewing the demonstration while preventing any unintended motion of the leaves by shielding them and causing the leaves to react only to charges which are intentionally brought to the leaves.
  • Metal case with glass windows
  • Includes instructions and foil
Specifications
Specifications
Instructions
Electroscope Metal Case with Foil
Glass, Metal