Supertek™ Electroscope, Simple DFS Item
This simple electroscope is a conical flask with rubber stopper which holds a metal rod with disc top and lower flat end to which a leaf may be pasted.
Supplier:  Supertek™ PH45246
Catalog No. S012323
Explore available promotions
Description
Description
  • Glass, conical flask with rubber stopper.
  • Rubber stopper holds a metal rod with disc top.
  • The rods lower flat end to which a leaf may be pasted.
  • Supplied with a booklet of 6 leaves.
  • Includes instructions
Specifications
Specifications
Electroscope Kit