Mopec Elevating Center Approach Wall Mount Autopsy Sink
Designed to provide an adjustable working area ergonomically correct for users of various heights. Mopec™ Elevating Center Approach Wall Mount Autopsy Sink can be surface mounted or recessed within the thickness of the wall.
Supplier:  Mopec CC200
Catalog No. 22444016
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Stainless steel construction
  • Center sink approach
  • Heavy duty elevating frame structure with dual synchronized actuators and push button controls
  • Hydro aspirator with reversing flow valve
  • Includes hose hanger and 8 foot of clear PVC tubing
  • Two cold water body rinse valve with vacuum breaker and serrated hose fitting
  • Two hot and cold water fixture with wrist blade handles, 8 in. swing spout with vacuum breaker
  • Four interchangeable removable grid plates stainless steel construction with .5 in. recess and .375 in. diameter holes on 2 in. square centers
  • Two GFCI duplex outlet with waterproof cover
  • Two instrument drawers with recessed handle and drop in removable pan
Warranty
1 Year Warranty
Specifications
Specifications
Autopsy Sink
Two GFCI Duplex Outlet with Waterproof Cover
80.01 cm
58.5 in. to 66.5 in.
289.56 cm
Stainless Steel
31.5 in.
114 in.
148.59 cm to 168.91 cm