Invitrogen™ eBioscience™ ELISA Wash Buffer, 1L packets
Suitable for washing ELISA plate microwells. eBioscience™ ELISA Wash Buffer is supplied in 10 packets of concentrated powder, yielding a total of 10 liters of 1x Wash Buffer.
Supplier:  Invitrogen™ 00040046
Catalog No. 5018479
Explore available promotions
Description
Description
The eBioscience ELISA Wash Buffer Powder is a dried powder formulation of phosphate buffered saline with 0.05% Tween-20. Each packet is reconstituted in 1 Liter of distilled, deionized water, resulting in 1x PBS/Tween-20 buffer (pH 7.4).
Specifications
Specifications
Wash Buffer
Powder
eBioscience™
10 Packets
ELISA
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.