missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ellagic Acid Dihydrate 98.0+%, TCI America™

Supplier:  TCI America E037525G

 View more versions of this product

Catalog No. E037525G


Explore available promotions

Description

Description

Specifications

Specifications

Ellagic Acid Dihydrate
450°C
MFCD00149494
2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione
O.O.OC1=C(O)C2=C3C(=C1)C(=O)OC1=C3C(=CC(O)=C1O)C(=O)O2
338.22
302.19
Crystalline Powder
133039-73-3
C14H10O10
25 g
ZEPCRIPMALGRJR-UHFFFAOYSA-N
6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4,6,8(16),11,13-hexaene-3,10-dione dihydrate
16760409
≥98.0% (HPLC,T)

Chemical Identifiers

133039-73-3
338.22
ZEPCRIPMALGRJR-UHFFFAOYSA-N
16760409
O.O.OC1=C(O)C2=C3C(=C1)C(=O)OC1=C3C(=CC(O)=C1O)C(=O)O2
C14H10O10
MFCD00149494
2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione
6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4,6,8(16),11,13-hexaene-3,10-dione dihydrate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DJ2620000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions