missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ellagic Acid Dihydrate 98.0+%, TCI America™
Supplier: TCI America E03755G
Specifications
Ellagic Acid Dihydrate | |
450°C | |
MFCD00149494 | |
2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione | |
O.O.OC1=C(O)C2=C3C(=C1)C(=O)OC1=C3C(=CC(O)=C1O)C(=O)O2 | |
338.22 | |
302.19 | |
Crystalline Powder |
133039-73-3 | |
C14H10O10 | |
5 g | |
ZEPCRIPMALGRJR-UHFFFAOYSA-N | |
6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0â´,¹â¶.0¹¹,¹âµ]hexadeca-1(15),4,6,8(16),11,13-hexaene-3,10-dione dihydrate | |
16760409 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
133039-73-3 | |
338.22 | |
ZEPCRIPMALGRJR-UHFFFAOYSA-N | |
16760409 | |
O.O.OC1=C(O)C2=C3C(=C1)C(=O)OC1=C3C(=CC(O)=C1O)C(=O)O2 |
C14H10O10 | |
MFCD00149494 | |
2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione | |
6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0â´,¹â¶.0¹¹,¹âµ]hexadeca-1(15),4,6,8(16),11,13-hexaene-3,10-dione dihydrate |
Safety and Handling
RTECSNumber : DJ2620000
TSCA : No