Agilent BioTek ELx405 Select Deep Well Microplate Washer
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates.
Supplier:  Agilent BioTek ELX405CVSSN
 View more versions of this product
Catalog No. EX405UCWVSD
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. The ELx405 Select is available in several configurations to address a wide variety of plate washing applications, including ELISA and cell-based assays.
Maintenance is easy with the available patented Ultrasonic Advantage that automates and cleans manifold tubes to remove built-up salt and protein.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Certifications
CE and ETL marked
Specifications
Specifications
CE and TUV marked. RoHS compliant.
≤2 μL/well
Microplate Washer
1 to 10
One positive displacement pump
50/60 Hz
59°F to 86°F
96-/384-well manifold; Deep well; Ultrasonic Advantage; Automatic 4-Buffer Switching; Optimized cell washing for loosely adherent monolayers
35.6 cm
25.4 cm
43.2 cm
100/240 V
≤3% CV
Programmable in minutes and seconds, up to 60 min.
13.5 kg
Backlit LCD
10 to 80%, noncondensing
ELX405UCWVSD
15°C to 30°C
14 in.
10 in.
17 in.
30 lb.
<8 A