Learn More
Agilent BioTek ELx405 Select Microplate Washer, IVD
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. For In Vitro Diagnostic Use.
$32394.49 - $40407.33
Specifications
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
---|---|
Precision | ≤3% CV |
Residual Volume | ≤2 μL/well |
Soak Time | Programmable in minutes and seconds, up to 60 min. |
Type | Microplate Washer |
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
BTELX405SI
|
Agilent BioTek
ELX405SI |
ELX405 | 96-well manifold |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTELX405CSI
|
Agilent BioTek
ELX405CSSI |
ELX405CS | 96-well manifold; Ultrasonic Advantage |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTELX405CVI
|
Agilent BioTek
ELX405CVSI |
ELX405CV | 96-well manifold; Automatic 4-Buffer Switching |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTELX405VSI
|
Agilent BioTek
ELX405CVSSI |
ELX405CVS | 96-well manifold; Ultrasonic Advantage; Automatic 4-Buffer Switching |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. The ELx405 Select is available in several configurations to address a wide variety of plate washing applications, including ELISA and cell-based assays.
Maintenance is easy with the available patented Ultrasonic Advantage that automates and cleans manifold tubes to remove built-up salt and protein.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Specifications
CE and TUV marked. RoHS compliant. | |
≤2 μL/well | |
Microplate Washer | |
1 to 10 | |
One positive displacement pump | |
50/60 Hz | |
15°C to 30°C | |
35.6 cm | |
25.4 cm | |
43.2 cm | |
100/240 V |
≤3% CV | |
Programmable in minutes and seconds, up to 60 min. | |
13.5 kg | |
Backlit LCD | |
10 to 80%, noncondensing | |
59°F to 86°F | |
14 in. | |
10 in. | |
17 in. | |
30 lb. | |
<8 A |