missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Protein G, Peroxidase Conjugate
Purified recombinant protein G conjugated to peroxidase
Supplier: MilliporeSigma™ 539322500UG
Description
- Binds ≥99% to human IgG immobilized on a column
- Form: Liquid
- Formulation: In 10mM PBS, 0.5mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6
- Purity: Single band at 33.8 kd by SDS-PAGE
- Preservative: 0.02% Thimerosal
- Applications: ELISA, Immunoblotting (Western Blotting)
Specifications
Protein G, Peroxidase Conjugate | |
500 μg | |
2°C to 8°C |