missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Calbiochem™ Streptavidin, Phycoerythrin Conjugate
For Enzyme immunoassay and flow cytometry
Supplier: MilliporeSigma™ 1897371ML
Description
- Specific for biotin, biotinylated proteins, or biotinylated antibodies
- Form: Liquid
- Formulation: In 150mM NaCl, 50mM sodium phosphate, 1% BSA, 0.1% glycerol, pH 7.6. Ammonium sulfate free
- Preservative: ≤0.1% sodium azide
Specifications
Streptavidin, Phycoerythrin Conjugate | |
1 mL | |
2°C to 8°C |