MilliporeSigma™ Chemicon™ Characterization Kit
contains validated antibodies that characterize different developmental stages of oligodendrocytes along with two antibodies for the detection of neurons and astrocytes, respectively
Supplier: MilliporeSigma™ SCR601
Description
EMD Millipore's Human Oligodendrocyte Characterization Kit (SCR601) contains validated antibodies that characterize different developmental stages of oligodendrocytes along with two antibodies for the detection of neurons and astrocytes, respectively. The kit includes antibodies to NG2 Chondroitin Sulfate Proteoglycan, Myelin Proteolipid Protein, Galactocerebroside, Myelin Oligodendrocyte Glycoprotein, Glial Fibrillary Acidic Protein and MAP2. The kit provides a convenient way to characterize cell populations generated using EMD-Millipore's Human Oligodendrocyte Differentiation Kit (SCR601)Specifications
Rabbit anti-NG2 Chondroitin Sulfate Proteoglycan, 10μg; Mouse anti-Glial Fibrillary Acidic Protein, clone GA5, 10μg; Mouse anti-Myelin Oligodendrocyte Glycoprotein (MOG), 10μg; Mouse anti-Galactocerebroside, clone mGalC, 10μg; Mouse anti-Myelin Proteolipid Protein, C-terminus ,clone PLPC1 20μg | |
Cell Culture, Immunocytochemistry | |
1 Kit |