MilliporeSigma™ Chemicon™ FibroGRO™ Inactivated Xeno-Free Human Foreskin Fibroblasts
derived from normal human foreskin (male neonate) and have been isolated and manufactured under xeno-free conditions.
Supplier:  MilliporeSigma™ SCC057
Catalog No. SCC057
Explore available promotions
Description
Description
Millipore’s FibroGROTM Xeno-Free Human Foreskin Fibroblasts (Mitomycin C treated) are derived from normal human foreskin (male neonate) and have been isolated and manufactured under xeno-free conditions. Cells have been mitotically inactivated with Mitomycin C and validated not to replicate. Cells are supplied at a low passage number (p6 – p8), tested negative for mycoplasma and can be used as a feeder layer to maintain human embryonic stem cell culture in an undifferentiated state.
Specifications
Specifications
4 vials of Mit-C treated fibroblasts. Store in liquid nitrogen
Stem cell culture and cell culture
1 Kit
Human foreskin fibroblasts