MilliporeSigma™ Chemicon™ In vitro Angiogenesis Assay Kit
Activity Assay
Supplier: MilliporeSigma™ ECM625
Specifications
Store kit materials at -20°C for up to 6 months. Once ECMatrix has been thawed, 2-8°C for one week | |
Activity Assay | |
1 Kit |
In vitro angiogenesis assay kit | |
ECMatrix Diluent Buffer, 10X, ECMatrix Gel Solution |