missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ EmbryoMax™ Nucleosides (100X)
For stem cell culture applications.
Supplier: MilliporeSigma™ ES008D
Description
- Used for routine mouse embryonic stem cell culture applications
Specifications
Store at -20°C upto 1 year. | |
Nucleosides | |
Stem cell culture |
Liquid | |
Cytidine: 0.73g/L; Guanosine: 0.85g/L; Uridine: 0.73g/L; Adenosine: 0.8g/L; Thymidine: 0.24g/L | |
50 mL |