MilliporeSigma™ EndoGRO™ Human Umbilical Vein Endothelial Cells (HUVEC)
a commonly studied human cell type extracted from human neonatal umbilical cords.
Supplier: MilliporeSigma™ SCCE001
Description
EndoGRO Human Umbilical Vein Endothelial Cells (HUVEC) are a commonly studied human cell type extracted from human neonatal umbilical cords. They are relatively easy to culture, and provide a valuable cell model for many vascular biology research applications, including inflammation, angiogenesis, atherosclerosis, blood clotting, vasoconstriction, and vasodilation. EndoGRO™ HUVEC are provided at passage 1, and have been quality tested in low-serum media without exposure to phenol red.Specifications
Cell Culture | |
500,000 cells | |
Human neonatal umbilical cords |