missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ EZ-Fit™ Filtration Unit

For bioburden testing of liquid samples like water, process samples, or final products

Supplier:  MilliporeSigma™ EFHAW100B

 View more versions of this product

Catalog No. EFHAW100B


Explore available promotions

Description

Description

  • Designed for optimizing and securing laboratory workflow to provide time-saving and reliable microbiological results
  • Comprise of 100mL or 250mL funnels
  • Black or white gridded membranes
  • After filtration of sample, membrane can be transferred and cultured on agar plate
  • Sterilized and disposable

Conform to international standards (EP/USP) and comply with water testing regulations.

Specifications

Specifications

Black with White Grid
0.45 μm
Bulk Packed
48/Pk.
100 mL
MCE
Sterile
48
Filtration Unit
Videos
SDS
Documents

Documents

Promotions

Promotions