MilliporeSigma™ Flow-limiting Orifices
Creates quantitative airflow through a filter for airborne contamination analysis
Supplier: MilliporeSigma™ XX5000002
Description
Flow Limiting Orifices create quantitative airflow through a MF-Millipore filter for airborne contamination analysis. The orifices are threaded into the outlet of a Millipore Aerosol Analysis Holder or Aerosol Adapter to enable a constant flow rate when required vacuum level is applied.Ratings are accurate to within approximately 15%. For greater precision, calibrate orifice with a flowmeter at actual working vacuum level.Specifications
Environmental Test Kits or Supplies | |
Millipore Aerosol Analysis Holder or Aerosol Adapter | |
1 L/min. |
Stainless Steel | |
To create quantitative airflow through a Millipore filter for airborne contamination analysis | |
Flow-limiting Orifice, 1 L/min |