MilliporeSigma™ Histosec™ Pastilles (without DMSO) Embedding Agent for Histology
solidification point 56-58°C, embedding agent for histology
$643.11 - $1234.24
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M1151619025
|
MilliporeSigma™
1151619025 |
25kg |
Each for $1,234.24
|
|
|||||
M1151612504
|
MilliporeSigma™
1151612504 |
4 x 2.5kg |
Each for $643.11
|
|
|||||
Description
Histosec™ pastilles (without DMSO) is an embedding medium for histology of samples of human origin. It is a paraffin enriched with polymers, also available with the addition of DMSO (dimethyl sulfoxide). DMSO increases the rate of penetration of paraffin and provides additional preservation of the tissue structures. Its melting range is between 56 - 58 °C and, within this range any specimen can be processed without fear of damage irrespective of the method used.Specifications
Pastilles |